Learn More
Biomatik Corporation Somatostatin (SST) anti-Mouse, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU27583
Encompass_Preferred
Somatostatin (Sst) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Somatostatin (SST). The Somatostatin (Sst) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Ser27~Cys116. Applications: WB, IHC, ICC, IP. Target Synonyms: Somatostatin (SST). Accession Number: P60041. Gene ID: 20604. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9982