Learn More
Biomatik Corporation Somatostatin (SST) anti-Human, Rat, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU27584
Encompass_Preferred
Somatostatin (Sst) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Somatostatin (SST). The Somatostatin (Sst) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Met1~Cys116. Applications: WB, IHC, ICC, IP. Target Synonyms: Somatostatin (SST). Accession Number: P61278. Gene ID: 6750. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9983