Learn More
Biomatik Corporation Cyclin D1 (CCND1) anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501509987
Encompass_Preferred
The Cyclin D1 (CCND1) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse, Rat. Immunogen: CCND1 (Met1-Ile295). Applications: WB, IHC. Alternative Names: CCN-D1; BCL1; PRAD1; U21B31; G1/S-Specific Cyclin-D1; Parathyroid Adenomatosis 1; B-Cell CLL/Lymphoma 1; G1/S-Specific Cyclin D1; PRAD1 oncogene. UniProt: P24385. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-9987 Supplier Biomatik Corporation Supplier No. CAU27596
The Cyclin D1 (CCND1) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse, Rat. Immunogen: CCND1 (Met1-Ile295). Applications: WB, IHC. Alternative Names: CCN-D1; BCL1; PRAD1; U21B31; G1/S-Specific Cyclin-D1; Parathyroid Adenomatosis 1; B-Cell CLL/Lymphoma 1; G1/S-Specific Cyclin D1; PRAD1 oncogene. UniProt: P24385. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.