Learn More
Biomatik Corporation S100 Calcium Binding Protein B (S100B) anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501509990
Encompass_Preferred
S100 Calcium Binding Protein B (S100B) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to S100 Calcium Binding Protein B (S100B). The S100 Calcium Binding Protein B (S100B) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Met1~Glu92. Applications: WB, IHC, ICC, IP. Target Synonyms: S100 Calcium Binding Protein B (S100B). Accession Number: P04271. Gene ID: 6285. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9990 Supplier Biomatik Corporation Supplier No. CAU27634
S100 Calcium Binding Protein B (S100B) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to S100 Calcium Binding Protein B (S100B). The S100 Calcium Binding Protein B (S100B) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Met1~Glu92. Applications: WB, IHC, ICC, IP. Target Synonyms: S100 Calcium Binding Protein B (S100B). Accession Number: P04271. Gene ID: 6285. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.