Learn More
Biomatik Corporation Interleukin 1 Beta (IL1b) anti-Bovine, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501509991
Encompass_Preferred
Interleukin 1 Beta (Il1B) Polyclonal Antibody is a Bovine (Bos taurus) Polyclonal Antibody to Interleukin 1 Beta (IL1b). The Interleukin 1 Beta (Il1B) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Bovine (Bos taurus). Immunogen: Met1~Pro266. Applications: WB, IHC, ICC, IP. Target Synonyms: Interleukin 1 Beta (IL1b). Accession Number: P09428. Gene ID: 281251. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9991 Supplier Biomatik Corporation Supplier No. CAU27649
Interleukin 1 Beta (Il1B) Polyclonal Antibody is a Bovine (Bos taurus) Polyclonal Antibody to Interleukin 1 Beta (IL1b). The Interleukin 1 Beta (Il1B) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Bovine (Bos taurus). Immunogen: Met1~Pro266. Applications: WB, IHC, ICC, IP. Target Synonyms: Interleukin 1 Beta (IL1b). Accession Number: P09428. Gene ID: 281251. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.