Learn More
Biomatik Corporation Intercellular Adhesion Molecule 1 (ICAM1) anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501509995
Encompass_Preferred
Intercellular Adhesion Molecule 1 (Icam1) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Intercellular Adhesion Molecule 1 (ICAM1). The Intercellular Adhesion Molecule 1 (Icam1) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Gly41~Gly464. Applications: WB, IHC, ICC, IP. Target Synonyms: Intercellular Adhesion Molecule 1 (ICAM1). Accession Number: P05362. Gene ID: 3383. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9995 Supplier Biomatik Corporation Supplier No. CAU27700
Intercellular Adhesion Molecule 1 (Icam1) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Intercellular Adhesion Molecule 1 (ICAM1). The Intercellular Adhesion Molecule 1 (Icam1) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Gly41~Gly464. Applications: WB, IHC, ICC, IP. Target Synonyms: Intercellular Adhesion Molecule 1 (ICAM1). Accession Number: P05362. Gene ID: 3383. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.