Learn More
Biomatik Corporation N-Terminal Pro-Atrial Natriuretic Peptide (NT-ProANP) anti-Human, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU27814
Encompass_Preferred
N-Terminal Pro-Atrial Natriuretic Peptide (Nt-Proanp) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to N-Terminal Pro-Atrial Natriuretic Peptide (NT-ProANP). The N-Terminal Pro-Atrial Natriuretic Peptide (Nt-Proanp) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Ala25~Arg123. Applications: WB, IHC, ICC, IP. Target Synonyms: N-Terminal Pro-Atrial Natriuretic Peptide (NT-ProANP). Accession Number: P01160. Gene ID: 4878. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0001