Learn More
Biomatik Corporation Osteocalcin (OC) anti-Human, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501510004
Encompass_Preferred
Osteocalcin (Oc) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Osteocalcin (OC). The Osteocalcin (Oc) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Ala21~Pro99. Applications: WB, IHC, ICC, IP. Target Synonyms: Osteocalcin (OC). Accession Number: P02818. Gene ID: 632. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0004 Supplier Biomatik Corporation Supplier No. CAU27847
Osteocalcin (Oc) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Osteocalcin (OC). The Osteocalcin (Oc) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Ala21~Pro99. Applications: WB, IHC, ICC, IP. Target Synonyms: Osteocalcin (OC). Accession Number: P02818. Gene ID: 632. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.