Learn More
Biomatik Corporation Androstenedione (ASD) anti-General, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU27866
Encompass_Preferred
The Androstenedione (ASD) Polyclonal Antibody is raised in Rabbit. Species Reactivity: General. Immunogen: Small Molecule, ASD conjugated to OVA. Applications: WB, IHC, ICC, IP. Alternative Names: 4-Androstenedione; 4-Androstene-3,17-Dione. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-151-0008