Learn More
Biomatik Corporation Insulin (INS) anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501510010
Encompass_Preferred
Insulin (Ins) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Insulin (INS). The Insulin (Ins) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Phe25~Asn110. Applications: WB, IHC, ICC, IP. Target Synonyms: Insulin (INS). Accession Number: P01308. Gene ID: 3630. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0010 Supplier Biomatik Corporation Supplier No. CAU27884
Insulin (Ins) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Insulin (INS). The Insulin (Ins) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Phe25~Asn110. Applications: WB, IHC, ICC, IP. Target Synonyms: Insulin (INS). Accession Number: P01308. Gene ID: 3630. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.