Learn More
Biomatik Corporation Octamer Binding Transcription Factor 4 (OCT4) anti-Human, Pig, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501510015
Encompass_Preferred
Octamer Binding Transcription Factor 4 (Oct4) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Octamer Binding Transcription Factor 4 (OCT4). The Octamer Binding Transcription Factor 4 (Oct4) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Ser136~Asn360. Applications: WB, IHC, ICC, IP. Target Synonyms: Octamer Binding Transcription Factor 4 (OCT4). Accession Number: Q01860. Gene ID: 5460. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0015 Supplier Biomatik Corporation Supplier No. CAU27923
Octamer Binding Transcription Factor 4 (Oct4) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Octamer Binding Transcription Factor 4 (OCT4). The Octamer Binding Transcription Factor 4 (Oct4) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Ser136~Asn360. Applications: WB, IHC, ICC, IP. Target Synonyms: Octamer Binding Transcription Factor 4 (OCT4). Accession Number: Q01860. Gene ID: 5460. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.