Learn More
Biomatik Corporation Sex Determining Region Y Box Protein 2 (SOX2) anti-Human, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501510018
Encompass_Preferred
Sex Determining Region Y Box Protein 2 (Sox2) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Sex Determining Region Y Box Protein 2 (SOX2). The Sex Determining Region Y Box Protein 2 (Sox2) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: His105~Met317. Applications: WB, IHC, ICC, IP. Target Synonyms: Sex Determining Region Y Box Protein 2 (SOX2). Accession Number: P48431. Gene ID: 6657. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0018 Supplier Biomatik Corporation Supplier No. CAU27954
Sex Determining Region Y Box Protein 2 (Sox2) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Sex Determining Region Y Box Protein 2 (SOX2). The Sex Determining Region Y Box Protein 2 (Sox2) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: His105~Met317. Applications: WB, IHC, ICC, IP. Target Synonyms: Sex Determining Region Y Box Protein 2 (SOX2). Accession Number: P48431. Gene ID: 6657. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.