Learn More
Biomatik Corporation Mucin 17 (MUC17) anti-Human, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501510019
Encompass_Preferred
Mucin 17 (Muc17) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Mucin 17 (MUC17). The Mucin 17 (Muc17) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Thr3773~Thr3998. Applications: WB, IHC, ICC, IP. Target Synonyms: Mucin 17 (MUC17). Accession Number: Q685J3. Gene ID: 140453. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0019 Supplier Biomatik Corporation Supplier No. CAU27955
Mucin 17 (Muc17) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Mucin 17 (MUC17). The Mucin 17 (Muc17) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Thr3773~Thr3998. Applications: WB, IHC, ICC, IP. Target Synonyms: Mucin 17 (MUC17). Accession Number: Q685J3. Gene ID: 140453. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.