Learn More
Biomatik Corporation Retinol Binding Protein 1, Cellular (RBP1) anti-Human, Rat, Pig, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU27966
Encompass_Preferred
Retinol Binding Protein 1, Cellular (Rbp1) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Retinol Binding Protein 1, Cellular (RBP1). The Retinol Binding Protein 1, Cellular (Rbp1) Polyclonal Antibody has been validated for the following applications: WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Pro64~Gln197. Applications: WB, IHC. Target Synonyms: Retinol Binding Protein 1, Cellular (RBP1). Accession Number: P09455. Gene ID: 5947. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0020