Learn More
Biomatik Corporation Dehydroepiandrosterone (DHEA) anti-General, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU27972
Encompass_Preferred
The Dehydroepiandrosterone (DHEA) Polyclonal Antibody is raised in Rabbit. Species Reactivity: General. Immunogen: Small Molecule, dehydroepiandrosterone conjugated to OVA. Applications: WB, IHC, ICC, IP. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-151-0021