Learn More
Biomatik Corporation Transforming Growth Factor Beta Receptor I (TGFbR1) anti-Human, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501510022
Encompass_Preferred
Transforming Growth Factor Beta Receptor I (Tgfbr1) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Transforming Growth Factor Beta Receptor I (TGFbR1). The Transforming Growth Factor Beta Receptor I (Tgfbr1) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Glu162~Ala403. Applications: WB, IHC, ICC, IP. Target Synonyms: Transforming Growth Factor Beta Receptor I (TGFbR1). Accession Number: P36897. Gene ID: 7046. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0022 Supplier Biomatik Corporation Supplier No. CAU27973
Transforming Growth Factor Beta Receptor I (Tgfbr1) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Transforming Growth Factor Beta Receptor I (TGFbR1). The Transforming Growth Factor Beta Receptor I (Tgfbr1) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Glu162~Ala403. Applications: WB, IHC, ICC, IP. Target Synonyms: Transforming Growth Factor Beta Receptor I (TGFbR1). Accession Number: P36897. Gene ID: 7046. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.