Learn More
Biomatik Corporation Interferon Gamma Induced Protein 10kDa (IP10) anti-Human, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU28036
Encompass_Preferred
Interferon Gamma Induced Protein 10Kda (Ip10) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Interferon Gamma Induced Protein 10kDa (IP10). The Interferon Gamma Induced Protein 10Kda (Ip10) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Val22~Pro98. Applications: WB, IHC, ICC, IP. Target Synonyms: Interferon Gamma Induced Protein 10kDa (IP10). Accession Number: P02778. Gene ID: 3627. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0027