Learn More
Biomatik Corporation Poly ADP Ribose Polymerase (PARP) anti-Human, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501510036
Encompass_Preferred
Poly Adp Ribose Polymerase (Parp) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Poly ADP Ribose Polymerase (PARP). The Poly Adp Ribose Polymerase (Parp) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Asp788~Trp1014. Applications: WB, IHC, ICC, IP. Target Synonyms: Poly ADP Ribose Polymerase (PARP). Accession Number: P09874. Gene ID: 142. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0036 Supplier Biomatik Corporation Supplier No. CAU28202
Poly Adp Ribose Polymerase (Parp) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Poly ADP Ribose Polymerase (PARP). The Poly Adp Ribose Polymerase (Parp) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Asp788~Trp1014. Applications: WB, IHC, ICC, IP. Target Synonyms: Poly ADP Ribose Polymerase (PARP). Accession Number: P09874. Gene ID: 142. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.