Learn More
Biomatik Corporation Connexin 43 (CX43) anti-Human, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501510038
Encompass_Preferred
The Connexin 43 (CX43) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: CX43 (Ser180-Ile382). Applications: WB, IHC. Alternative Names: ODDD; GJAL; GJA1; GJ-A1; Gap junction 43 kDa heart protein; Gap Junction Protein Alpha 1; Oculodentodigital Dysplasia; Syndactyly Type III. UniProt: P17302. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-151-0038 Supplier Biomatik Corporation Supplier No. CAU28209
The Connexin 43 (CX43) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: CX43 (Ser180-Ile382). Applications: WB, IHC. Alternative Names: ODDD; GJAL; GJA1; GJ-A1; Gap junction 43 kDa heart protein; Gap Junction Protein Alpha 1; Oculodentodigital Dysplasia; Syndactyly Type III. UniProt: P17302. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.