Learn More
Biomatik Corporation Interleukin 1 Receptor Antagonist (IL1RA) anti-Bovine, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU28310
Encompass_Preferred
Interleukin 1 Receptor Antagonist (Il1Ra) Polyclonal Antibody is a Bovine (Bos taurus) Polyclonal Antibody to Interleukin 1 Receptor Antagonist (IL1RA). The Interleukin 1 Receptor Antagonist (Il1Ra) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Bovine (Bos taurus). Immunogen: His24~Asp174. Applications: WB, IHC, ICC, IP. Target Synonyms: Interleukin 1 Receptor Antagonist (IL1RA). Accession Number: O77482. Gene ID: 281860. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0042