Learn More
Biomatik Corporation Interferon Beta (IFNb) anti-Dog, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU28316
Encompass_Preferred
Interferon Beta (Ifnb) Polyclonal Antibody is a Dog (Canis familiaris; Canine) Polyclonal Antibody to Interferon Beta (IFNb). The Interferon Beta (Ifnb) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Dog (Canis familiaris; Canine). Immunogen: Ala21~Asn186. Applications: WB, IHC, ICC, IP. Target Synonyms: Interferon Beta (IFNb). Accession Number: B6E116. Gene ID: 481558. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0043