Learn More
Biomatik Corporation Alpha-1-Microglobulin/Bikunin Precursor (a1M) anti-Mouse, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501510045
Encompass_Preferred
The Alpha-1-Microglobulin/Bikunin Precursor (a1M) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Mouse. Immunogen: a1M (Arg18-Ala202). Applications: WB, IHC. Alternative Names: AMBP; UTI; HCP; EDC1; HI30; IATIL; ITILC; ITI; ITIL; Alpha 1 Microglobulin/Bikunin Precursor; Growth-inhibiting protein 19; Uristatin; Uronic-Acid-Rich Protein; Trypstatin. UniProt: Q07456. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-151-0045 Supplier Biomatik Corporation Supplier No. CAU28334
The Alpha-1-Microglobulin/Bikunin Precursor (a1M) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Mouse. Immunogen: a1M (Arg18-Ala202). Applications: WB, IHC. Alternative Names: AMBP; UTI; HCP; EDC1; HI30; IATIL; ITILC; ITI; ITIL; Alpha 1 Microglobulin/Bikunin Precursor; Growth-inhibiting protein 19; Uristatin; Uronic-Acid-Rich Protein; Trypstatin. UniProt: Q07456. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.