Learn More
Biomatik Corporation Alanine Aminotransferase (ALT) anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501510047
Encompass_Preferred
The Alanine Aminotransferase (ALT) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse, Rat. Immunogen: ALT (Ile147-Phe263). Applications: WB, IHC. Alternative Names: GPT; ALAT; SGPT; AAT1; GPT1; Serum Glutamic Pyruvic Transaminase; Alanine Transaminase; Glutamate pyruvate transaminase 1; Glutamic--alanine transaminase 1. UniProt: P24298. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-151-0047 Supplier Biomatik Corporation Supplier No. CAU28356
The Alanine Aminotransferase (ALT) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse, Rat. Immunogen: ALT (Ile147-Phe263). Applications: WB, IHC. Alternative Names: GPT; ALAT; SGPT; AAT1; GPT1; Serum Glutamic Pyruvic Transaminase; Alanine Transaminase; Glutamate pyruvate transaminase 1; Glutamic--alanine transaminase 1. UniProt: P24298. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.