Learn More
Biomatik Corporation Growth Arrest Specific Protein 6 (GAS6) anti-Mouse, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU28362
Encompass_Preferred
The Growth Arrest Specific Protein 6 (GAS6) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Mouse. Immunogen: GAS6 (Ala483-Gly646). Applications: WB, IHC. Alternative Names: AXLLG; AXSF; AXL Stimulatory Factor; AXL receptor tyrosine kinase ligand. UniProt: Q61592. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-151-0048