Learn More
Biomatik Corporation Galactosidase Alpha (GLa) anti-Mouse, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501510050
Encompass_Preferred
The Galactosidase Alpha (GLa) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Mouse. Immunogen: GLa (Val29-Gly258). Applications: WB, IHC. Alternative Names: GALA; GL-A; Alpha-D-Galactoside Galactohydrolase; Alpha-D-Galactosidase A; Melibiase; Agalsidase. UniProt: P51569. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-151-0050 Supplier Biomatik Corporation Supplier No. CAU28385
The Galactosidase Alpha (GLa) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Mouse. Immunogen: GLa (Val29-Gly258). Applications: WB, IHC. Alternative Names: GALA; GL-A; Alpha-D-Galactoside Galactohydrolase; Alpha-D-Galactosidase A; Melibiase; Agalsidase. UniProt: P51569. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.