Learn More
Biomatik Corporation Ubiquitin (Ub) anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU28423
Encompass_Preferred
Ubiquitin (Ub) Polyclonal Antibody is a Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus), Rabbit (Oryctolagus cuniculus), Dog (Canis familiaris; Canine), Pig (Sus scrofa; Porcine), Bovine (Bos taurus), Horse (Equus caballus; Equine) Polyclonal Antibody to Ubiquitin (Ub). The Ubiquitin (Ub) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus), Rabbit (Oryctolagus cuniculus), Dog (Canis familiaris; Canine), Pig (Sus scrofa; Porcine), Bovine (Bos taurus), Horse (Equus caballus; Equine). Immunogen: Met1~Cys229. Applications: WB, IHC, ICC, IP. Target Synonyms: Ubiquitin (Ub). Accession Number: P0CG47. Gene ID: 7314. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0056