Learn More
Biomatik Corporation Carbohydrate Antigen 125 (CA125) anti-Mouse, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU28435
Encompass_Preferred
The Carbohydrate Antigen 125 (CA125) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Mouse. Immunogen: CA125 (Thr2-Gln258). Applications: WB. Alternative Names: MUC16; Cancer Antigen 125; Mucin 16,Cell Surface Associated; Ovarian Cancer-Related Tumor Marker CA125; Ovarian Carcinoma Antigen CA125. UniProt: core/X. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-151-0058