Learn More
Biomatik Corporation Alpha-Fetoprotein (aFP) anti-Human, Pig, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU28443
Encompass_Preferred
The Alpha-Fetoprotein (aFP) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Pig. Immunogen: aFP (Ile31-Pro171). Applications: WB, IHC. Alternative Names: A-FP; FETA; HPAFP; Alpha-Fetoglobulin; Alpha-1-fetoprotein. UniProt: P27435. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-151-0059