Learn More
Biomatik Corporation Vascular Endothelial Growth Factor A (VEGFA) anti-Human, Rat, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501510061
Encompass_Preferred
Vascular Endothelial Growth Factor A (Vegfa) Polyclonal Antibody is a Rat (Rattus norvegicus) Polyclonal Antibody to Vascular Endothelial Growth Factor A (VEGFA). The Vascular Endothelial Growth Factor A (Vegfa) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Rat (Rattus norvegicus). Immunogen: Ala27~Arg190. Applications: WB, IHC, ICC, IP. Target Synonyms: Vascular Endothelial Growth Factor A (VEGFA). Accession Number: P16612. Gene ID: 83785. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0061 Supplier Biomatik Corporation Supplier No. CAU28469
Vascular Endothelial Growth Factor A (Vegfa) Polyclonal Antibody is a Rat (Rattus norvegicus) Polyclonal Antibody to Vascular Endothelial Growth Factor A (VEGFA). The Vascular Endothelial Growth Factor A (Vegfa) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Rat (Rattus norvegicus). Immunogen: Ala27~Arg190. Applications: WB, IHC, ICC, IP. Target Synonyms: Vascular Endothelial Growth Factor A (VEGFA). Accession Number: P16612. Gene ID: 83785. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.