Learn More
Biomatik Corporation Vascular Endothelial Growth Factor A (VEGFA) anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU28471
Encompass_Preferred
Vascular Endothelial Growth Factor A (Vegfa) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Vascular Endothelial Growth Factor A (VEGFA). The Vascular Endothelial Growth Factor A (Vegfa) Polyclonal Antibody has been validated for the following applications: WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Pro28~Arg147. Applications: WB, IHC. Target Synonyms: Vascular Endothelial Growth Factor A (VEGFA). Accession Number: P15692. Gene ID: 7422. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0062