Learn More
Biomatik Corporation Tumor Necrosis Factor Alpha (TNFa) anti-Human, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501510066
Encompass_Preferred
Tumor Necrosis Factor Alpha (Tnfa) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Tumor Necrosis Factor Alpha (TNFa). The Tumor Necrosis Factor Alpha (Tnfa) Polyclonal Antibody has been validated for the following applications: WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Val77~Leu233. Applications: WB, IHC, IF. Target Synonyms: Tumor Necrosis Factor Alpha (TNFa). Accession Number: P01375. Gene ID: 7124. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0066 Supplier Biomatik Corporation Supplier No. CAU28502
Tumor Necrosis Factor Alpha (Tnfa) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Tumor Necrosis Factor Alpha (TNFa). The Tumor Necrosis Factor Alpha (Tnfa) Polyclonal Antibody has been validated for the following applications: WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Val77~Leu233. Applications: WB, IHC, IF. Target Synonyms: Tumor Necrosis Factor Alpha (TNFa). Accession Number: P01375. Gene ID: 7124. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.