Learn More
Biomatik Corporation TEK Tyrosine Kinase, Endothelial (Tie2) anti-Human, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU28529
Encompass_Preferred
Tek Tyrosine Kinase, Endothelial (Tie2) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TEK Tyrosine Kinase, Endothelial (Tie2). The Tek Tyrosine Kinase, Endothelial (Tie2) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Gly286~Ile465. Applications: WB, IHC, ICC, IP. Target Synonyms: TEK Tyrosine Kinase, Endothelial (Tie2). Accession Number: Q02763. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0070