Learn More
Biomatik Corporation Transforming Growth Factor Beta 1 (TGFb1) anti-Mouse, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU28537
Encompass_Preferred
Transforming Growth Factor Beta 1 (Tgfb1) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Transforming Growth Factor Beta 1 (TGFb1). The Transforming Growth Factor Beta 1 (Tgfb1) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Ala279~Ser390. Applications: WB, IHC, ICC, IP. Target Synonyms: Transforming Growth Factor Beta 1 (TGFb1). Accession Number: P04202. Gene ID: 21803. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0072