Learn More
Biomatik Corporation Placenta Growth Factor (PLGF) anti-Rat, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501510075
Encompass_Preferred
Placenta Growth Factor (Plgf) Polyclonal Antibody is a Rat (Rattus norvegicus) Polyclonal Antibody to Placenta Growth Factor (PLGF). The Placenta Growth Factor (Plgf) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Rat (Rattus norvegicus). Immunogen: Ala24~Leu158. Applications: WB, IHC, ICC, IP. Target Synonyms: Placenta Growth Factor (PLGF). Accession Number: Q63434. Gene ID: 94203. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0075 Supplier Biomatik Corporation Supplier No. CAU28579
Placenta Growth Factor (Plgf) Polyclonal Antibody is a Rat (Rattus norvegicus) Polyclonal Antibody to Placenta Growth Factor (PLGF). The Placenta Growth Factor (Plgf) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Rat (Rattus norvegicus). Immunogen: Ala24~Leu158. Applications: WB, IHC, ICC, IP. Target Synonyms: Placenta Growth Factor (PLGF). Accession Number: Q63434. Gene ID: 94203. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.