Learn More
Biomatik Corporation Interleukin 2 (IL2) anti-Chicken, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU28728
Encompass_Preferred
Interleukin 2 (Il2) Polyclonal Antibody is a Chicken (Gallus) Polyclonal Antibody to Interleukin 2 (IL2). The Interleukin 2 (Il2) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Chicken (Gallus). Immunogen: Met1~Lys142. Applications: WB, IHC, ICC, IP. Target Synonyms: Interleukin 2 (IL2). Accession Number: Q6UG48. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0086