Learn More
Biomatik Corporation Defensin Beta 2 (DEFb2) anti-Human, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU28732
Encompass_Preferred
The Defensin Beta 2 (DEFb2) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: DEFb2 (Gly24-Pro64). Applications: WB, IHC. Alternative Names: B-DF2; DEFB2; BD2; DEF-B2; HBD2; DEFB102; HBD-2; SAP1; DEFB4A; DEFB4; Skin-Antimicrobial Peptide 1. UniProt: O15263. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-151-0088