Learn More
Biomatik Corporation Interleukin 18 (IL18) anti-Pig, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501510091
Encompass_Preferred
Interleukin 18 (Il18) Polyclonal Antibody is a Pig (Sus scrofa; Porcine) Polyclonal Antibody to Interleukin 18 (IL18). The Interleukin 18 (Il18) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Pig (Sus scrofa; Porcine). Immunogen: Tyr36~Asn192. Applications: WB, IHC, ICC, IP. Target Synonyms: Interleukin 18 (IL18). Accession Number: O19073. Gene ID: 397057. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0091 Supplier Biomatik Corporation Supplier No. CAU28767
Interleukin 18 (Il18) Polyclonal Antibody is a Pig (Sus scrofa; Porcine) Polyclonal Antibody to Interleukin 18 (IL18). The Interleukin 18 (Il18) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Pig (Sus scrofa; Porcine). Immunogen: Tyr36~Asn192. Applications: WB, IHC, ICC, IP. Target Synonyms: Interleukin 18 (IL18). Accession Number: O19073. Gene ID: 397057. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.