Learn More
Biomatik Corporation Interleukin 18 (IL18) anti-Chicken, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU28770
Encompass_Preferred
Interleukin 18 (Il18) Polyclonal Antibody is a Chicken (Gallus) Polyclonal Antibody to Interleukin 18 (IL18). The Interleukin 18 (Il18) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Chicken (Gallus). Immunogen: Met1~Leu196. Applications: WB, IHC, ICC, IP. Target Synonyms: Interleukin 18 (IL18). Accession Number: Q8QFQ8. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0093