Learn More
Biomatik Corporation Interleukin 10 (IL10) anti-Dog, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU28813
Encompass_Preferred
Interleukin 10 (Il10) Polyclonal Antibody is a Dog (Canis familiaris; Canine) Polyclonal Antibody to Interleukin 10 (IL10). The Interleukin 10 (Il10) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Dog (Canis familiaris; Canine). Immunogen: Arg21~Ile181. Applications: WB, IHC, ICC, IP. Target Synonyms: Interleukin 10 (IL10). Accession Number: P48411. Gene ID: 403628. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0099