Learn More
Biomatik Corporation Interferon Gamma (IFNg) anti-Human, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU28849
Encompass_Preferred
Interferon Gamma (Ifng) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Interferon Gamma (IFNg). The Interferon Gamma (Ifng) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Gln24~Gln166. Applications: WB, IHC, ICC, IP. Target Synonyms: Interferon Gamma (IFNg). Accession Number: P01579. Gene ID: 3458. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0102