Learn More
Biomatik Corporation Colony Stimulating Factor 2, Granulocyte Macrophage (GMCSF) anti-Human, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501510106
Encompass_Preferred
The Colony Stimulating Factor 2, Granulocyte Macrophage (GMCSF) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: GMCSF (Ala18-Glu144). Applications: WB. Alternative Names: CSF2; GM-CSF; Sargramostim; Molgramostin; Granulocyte-Macrophage Colony Stimulating Factor. UniProt: P04141. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-151-0106 Supplier Biomatik Corporation Supplier No. CAU28869
The Colony Stimulating Factor 2, Granulocyte Macrophage (GMCSF) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: GMCSF (Ala18-Glu144). Applications: WB. Alternative Names: CSF2; GM-CSF; Sargramostim; Molgramostin; Granulocyte-Macrophage Colony Stimulating Factor. UniProt: P04141. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.