Learn More
Biomatik Corporation Fibronectin (FN) anti-Mouse, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU28894
Encompass_Preferred
The Fibronectin (FN) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Mouse. Immunogen: FN (Gly2117-Leu2236). Applications: WB, IHC. Alternative Names: FN1; CIG; FINC; LETS; MSF; GFND2; Anastellin; Migration-Stimulating Factor; Cold-Insoluble Globulin; Large, External, Transformation-Sensitive Protein. UniProt: P11276. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-151-0107