Learn More
Biomatik Corporation Interferon Alpha (IFNa) anti-Dog, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501510111
Encompass_Preferred
Interferon Alpha (Ifna) Polyclonal Antibody is a Dog (Canis familiaris; Canine) Polyclonal Antibody to Interferon Alpha (IFNa). The Interferon Alpha (Ifna) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Dog (Canis familiaris; Canine). Immunogen: Ala23~Lys187. Applications: WB, IHC, ICC, IP. Target Synonyms: Interferon Alpha (IFNa). Accession Number: P81255. Gene ID: 100685213. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0111 Supplier Biomatik Corporation Supplier No. CAU28914
Interferon Alpha (Ifna) Polyclonal Antibody is a Dog (Canis familiaris; Canine) Polyclonal Antibody to Interferon Alpha (IFNa). The Interferon Alpha (Ifna) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Dog (Canis familiaris; Canine). Immunogen: Ala23~Lys187. Applications: WB, IHC, ICC, IP. Target Synonyms: Interferon Alpha (IFNa). Accession Number: P81255. Gene ID: 100685213. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.