Learn More
Biomatik Corporation Factor Related Apoptosis (FAS) anti-Human, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU28933
Encompass_Preferred
The Factor Related Apoptosis (FAS) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: FAS (Ser22-Ser172). Applications: WB, IHC. Alternative Names: CD95; ALPS1A; ALPS1-A; APO1; APT1; FAS1; FASTM; TNFRSF6; Fas Receptor; TNF Receptor Superfamily Member 6; Tumor Necrosis Factor Receptor Superfamily Member 6. UniProt: P25445. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-151-0112