Learn More
Biomatik Corporation 25-Hydroxyvitamin D3 (HVD3) Rabbit Polyclonal Antibody (100UG)
Supplier:  Biomatik Corporation CAU29077
Encompass_Preferred
The 25-Hydroxyvitamin D3 (HVD3) Polyclonal Antibody is raised in Rabbit. Species Reactivity: All species. Immunogen: OVA Conjugated 25-Hydroxyvitamin D3 (HVD3). Applications: WB, IHC, ICC, IP. Alternative Names: 25-HVD3; 25HVD3; Calcifediol; Calcidiol; H-VD3; 25-Hydroxycholecalciferol; 25-Hydroxy Vitamin D3; 25 (OH)D. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-151-0123