Learn More
Biomatik Corporation Bcl2 Associated X Protein (Bax) anti-Pig, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU29086
Encompass_Preferred
The Bcl2 Associated X Protein (Bax) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Pig. Immunogen: Bax (Met1-Gly192). Applications: WB. Alternative Names: BCL2L4; Bax Zeta; Apoptosis regulator BAX; Bcl-2-like protein 4. UniProt: F1RIQ4. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-151-0124