Learn More
Biomatik Corporation Collagen Type I (COL1) anti-Mouse, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501510125
Encompass_Preferred
The Collagen Type I (COL1) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Mouse. Immunogen: OVA conjugated Collagen Type I (COL1). Applications: WB, IHC, ICC, IP. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-151-0125 Supplier Biomatik Corporation Supplier No. CAU29088
The Collagen Type I (COL1) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Mouse. Immunogen: OVA conjugated Collagen Type I (COL1). Applications: WB, IHC, ICC, IP. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.