Learn More
Biomatik Corporation Interleukin 6 (IL6) anti-Rabbit, Guinea Polyclonal Antibody (100UG)
Supplier:  Biomatik Corporation CAU29090
Encompass_Preferred
Interleukin 6 (Il6) Polyclonal Antibody is a Rabbit (Oryctolagus cuniculus) Polyclonal Antibody to Interleukin 6 (IL6). The Interleukin 6 (Il6) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Guinea pig. Species: Rabbit (Oryctolagus cuniculus). Immunogen: Phe27~Thr241. Applications: WB, IHC, ICC, IP. Target Synonyms: Interleukin 6 (IL6). Accession Number: Q9MZR1. Gene ID: 100008733. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0127