Learn More
Biomatik Corporation Immunoglobulin G (IgG) anti-Human, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501510129
Encompass_Preferred
Immunoglobulin G (Igg) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Immunoglobulin G (IgG). The Immunoglobulin G (Igg) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Applications: WB, IHC, ICC, IP. Target Synonyms: Immunoglobulin G (IgG). Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0129 Supplier Biomatik Corporation Supplier No. CAU29096
Immunoglobulin G (Igg) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Immunoglobulin G (IgG). The Immunoglobulin G (Igg) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Applications: WB, IHC, ICC, IP. Target Synonyms: Immunoglobulin G (IgG). Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.