Learn More
Biomatik Corporation Thyrotropin Releasing Hormone (TRH) anti-Human, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU29111
Encompass_Preferred
Thyrotropin Releasing Hormone (Trh) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Thyrotropin Releasing Hormone (TRH). The Thyrotropin Releasing Hormone (Trh) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Applications: WB, IHC, ICC, IP. Target Synonyms: Thyrotropin Releasing Hormone (TRH). Accession Number: Q62361. Gene ID: 22044. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0131